Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-β-homovaline 98%

Supplier: AMBEED, INC
Warning

A118133-1G A118133-100MG A118133-250MG
103798-998EA 78 USD InStock
103798-998 103824-890 76995-596 76995-598
Fmoc-L-β-homovaline 98%
Fmoc-L-beta-homovaline
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₄
Boiling Pt: 563.9 °C (760 mm Hg)
Flash Pt: 294.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862853
CAS Number: 172695-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty