Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ganciclovir 98%

Supplier: AMBEED, INC
Danger

A200267-1G A200267-5G A200267-100G A200267-10G A200267-25MG A200267-1G A200267-5G A200267-500G
103817-184EA 8 USD InStock
103817-184 103847-812 76873-366 76873-368 76873-370 77085-872 77085-874 77671-774
Ganciclovir 98%
Ganciclovir
Precaución: for Research Use Only


Formula: C₉H₁₃N₅O₄
MW: 255.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00870588
CAS Number: 82410-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty