Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Gemcitabine 98%

Supplier: AMBEED, INC
Warning

A433642-1G A433642-100G A433642-100MG A433642-10G A433642-250MG A433642-50MG A433642-1G A433642-25G A433642-5G
103827-560EA 10 USD InStock
103827-560 103838-628 76804-306 76804-308 76804-310 76804-312 76804-314 76842-330 76842-332 76842-334
Gemcitabine 98%
Gemcitabine
Precaución: for Research Use Only


Formula: C₉H₁₁F₂N₃O₄
Boiling Pt: 482.7 °C (760 mm Hg)
Melting Pt: 168.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00869720
CAS Number: 95058-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty