Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

GNE-9605 98%

Supplier: AMBEED, INC
Warning

A369305-10MG A369305-1MG A369305-25MG A369305-2MG A369305-50MG A369305-5MG
103791-660EA 82 USD InStock
103791-660 103792-674 103792-676 103799-628 77240-220 77240-222 77240-224 77240-226 77240-228 77240-230
GNE-9605 98%
GNE-9605
Precaución: for Research Use Only


Formula: C₁₇H₂₀ClF₄N₇O
MW: 449.83 g/mol
Boiling Pt: 587.9±60.0 °C (760 mm Hg)
Flash Pt: 309.4 °C
Storage Temperature: Freezer
CAS Number: 1536200-31-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty