Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nω-Nitro-L-arginine 98%

Supplier: AMBEED, INC
Danger

A289915-5G A289915-25G A289915-10G A289915-500G A289915-5G
103834-992EA 6 USD InStock
103834-992 76837-574 77153-962 77153-964 77153-966 77305-572
Nω-Nitro-L-arginine 98%
Nomega-Nitro-L-arginine
Precaución: for Research Use Only


Formula: C₆H₁₃N₅O₄
Boiling Pt: 462.4 °C (760 mm Hg)
Melting Pt: 257 °C
Storage Temperature: Ambient
MDL Number: MFCD00007033
CAS Number: 2149-70-4
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty