Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-L-aspartate 98%

Supplier: AMBEED, INC
Warning

A164363-1G A164363-5G A164363-1G A164363-25G
103805-854EA 5 USD InStock
103805-854 103844-114 77320-278 77320-280
1-Benzyl-L-aspartate 98%
1-Benzyl-L-aspartate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₄
Boiling Pt: 391 °C (760 mm Hg)
Melting Pt: 176 °C
Flash Pt: 190.3 °C
Storage Temperature: Freezer
MDL Number: MFCD00063186
CAS Number: 7362-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty