Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-D-Asp(OMe)-OH 97%

Supplier: AMBEED, INC
Warning

A142711-1G A142711-100G A142711-10G A142711-1G A142711-25G A142711-5G
103803-204EA 18 USD InStock
103803-204 77109-876 77109-878 77316-444 77316-446 77316-448
H-D-Asp(OMe)-OH 97%
H-D-Asp(OMe)-OH
Precaución: for Research Use Only


Formula: C₅H₉NO₄
Boiling Pt: 301.7 °C (760 mm Hg)
Flash Pt: 136.3 °C
Storage Temperature: Ambient
MDL Number: MFCD06797583
CAS Number: 21394-81-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty