Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Ethylphenylalanate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A233812-5G A233812-10G A233812-5G
103850-670EA 7 USD InStock
103850-670 77237-940 77303-966
L-Ethylphenylalanate hydrochloride 97%
L-Ethylphenylalanate hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClNO₂
MW: 229.703 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02063363
CAS Number: 19881-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty