Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Amino-2-(4-chlorophenyl)acetic acid 95%

Supplier: AMBEED, INC
Warning

A121738-1G A121738-10G A121738-250MG A121738-25G A121738-1G A121738-5G
103800-126EA 32 USD InStock
103800-126 76688-136 76688-138 76688-140 77312-252 77312-254
(R)-2-Amino-2-(4-chlorophenyl)acetic acid 95%
(R)-2-Amino-2-(4-chlorophenyl)acetic acid
Precaución: for Research Use Only


Formula: C₈H₈ClNO₂
MW: 185.6 g/mol
Boiling Pt: 328.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03839815
CAS Number: 43189-37-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty