Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Proline benzyl ester hydrochloride 96%

Supplier: AMBEED, INC
Warning

A123446-1G A123446-100G A123446-10G A123446-25G A123446-1G A123446-5G
103800-404EA 6 USD InStock
103800-404 76971-676 76971-678 76971-680 77297-940 77312-524
D-Proline benzyl ester hydrochloride 96%
D-Proline benzyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₅NO₂
Boiling Pt: 309 °C (760 mm Hg)
Melting Pt: 143…145 °C
Flash Pt: 140.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153458
CAS Number: 53843-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty