Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Gln-OtBu·HCl 98%

Supplier: AMBEED, INC
Warning

A125895-1G A125895-250MG A125895-1G A125895-25G A125895-5G
103800-734EA 14 USD InStock
103800-734 76971-864 77312-890 77312-892 77312-894
H-Gln-OtBu·HCl 98%
H-Gln-OtBu·HCl
Precaución: for Research Use Only


Formula: C₉H₁₉ClN₂O₃
MW: 238.712 g/mol
Boiling Pt: 359.5 °C (760 mm Hg)
Flash Pt: 171.6 °C
Storage Temperature: Freezer
MDL Number: MFCD00039081
CAS Number: 39741-62-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty