Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Glutamic acid 5-methyl ester 97%

Supplier: AMBEED, INC
Warning

A157715-5G A157715-1000G A157715-100G A157715-10G A157715-500G A157715-5G A157715-25G
103843-242EA 6 USD InStock
103843-242 77273-182 77273-184 77273-186 77273-188 77300-286 77319-080
L-Glutamic acid 5-methyl ester 97%
L-Glutamic acid 5-methyl ester
Precaución: for Research Use Only


Formula: C₆H₁₁NO₄
MW: 161.2 g/mol
Boiling Pt: 316.1 °C (760 mm Hg)
Melting Pt: 182 °C
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00002632
CAS Number: 1499-55-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty