Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-Glu(OtBu)-NH₂·HCl 98%

Supplier: AMBEED, INC
Warning

A537862-1G A537862-25G A537862-5G A537862-100G A537862-10G A537862-250MG
103828-606EA 8 USD InStock
103828-606 103839-728 76843-762 76843-764 76843-766 77005-002 77005-004 77005-006
H-Glu(OtBu)-NH₂·HCl 98%
H-Glu(OtBu)-NH₂·HCl
Precaución: for Research Use Only


Formula: C₉H₁₉ClN₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00237067
CAS Number: 108607-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty