Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-1-Benzyl-5-tert-butyl-2-aminopentanedioate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A100578-1G A100578-5G A100578-10G A100578-250MG A100578-1G A100578-5G
103795-520EA 45 USD InStock
103795-520 103821-210 77160-162 77160-164 77282-516 77282-518
(S)-1-Benzyl-5-tert-butyl-2-aminopentanedioate hydrochloride 97%
(S)-1-Benzyl-5-tert-butyl-2-aminopentanedioate hydrochloride
Precaución: for Research Use Only


Formula: C₁₆H₂₄ClNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD16036702
CAS Number: 98102-32-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty