Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

H-HoLeu-OH 95%

Supplier: AMBEED, INC
Warning

A100924-1G A100924-5G A100924-100MG A100924-250MG A100924-1G A100924-5G
103795-702EA 39 USD InStock
103795-702 103821-390 76765-146 76765-148 77296-912 77296-914
H-HoLeu-OH 95%
H-HoLeu-OH
Precaución: for Research Use Only


Formula: C₇H₁₅NO₂
Melting Pt: 254…256 °C
Storage Temperature: Ambient
MDL Number: MFCD03788077
CAS Number: 31872-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty