Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nε-(tert-butoxycarbonyl)-L-lysine 97%

Supplier: AMBEED, INC
Warning

A104053-1G A104053-5G A104053-100G A104053-10G A104053-25G A104053-500G A104053-5G
103796-646EA 5 USD InStock
103796-646 103822-358 76994-710 76994-712 76994-714 76994-716 77283-190
Nε-(tert-butoxycarbonyl)-L-lysine 97%
Nepsilon-(tert-butoxycarbonyl)-L-lysine
Precaución: for Research Use Only


Formula: C₁₁H₂₂N₂O₄
MW: 246.31 g/mol
Boiling Pt: 412.9 °C (760 mm Hg)
Melting Pt: 250 °C
Flash Pt: 203.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00037221
CAS Number: 2418-95-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty