Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Phenylalanine ethyl ester hydrochloride 98%

Supplier: AMBEED, INC
Warning

A138312-100G A138312-500G A138312-25G A138312-5G
76798-126EA 18 USD InStock
76798-126 76798-128 76798-130 77298-978 77298-980
L-Phenylalanine ethyl ester hydrochloride 98%
L-Phenylalanine ethyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₆ClNO₂
Flash Pt: 140.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00012507
CAS Number: 3182-93-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty