Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Phenylalanine-tert-butyl ester hydrochloride 97%

Supplier: AMBEED, INC
Warning

A160004-1G A160004-5G A160004-100G A160004-10G A160004-500G A160004-1G A160004-5G A160004-25G
103805-314EA 6 USD InStock
103805-314 103843-544 76997-758 76997-760 76997-762 77300-430 77300-432 77319-450
L-Phenylalanine-tert-butyl ester hydrochloride 97%
L-Phenylalanine-tert-butyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₀ClNO₂
Melting Pt: 240 °C
Flash Pt: 158.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038894
CAS Number: 15100-75-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty