Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Serinamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A142497-100G A142497-10G A142497-25G A142497-5G
103803-166EA 8 USD InStock
103803-166 76811-830 76811-832 77316-390 77316-392
L-Serinamide hydrochloride 97%
L-Serinamide hydrochloride
Precaución: for Research Use Only


Formula: C₃H₉ClN₂O₂
MW: 140.567 g/mol
Boiling Pt: 444.7 °C (760 mm Hg)
Melting Pt: 185…188 °C
Flash Pt: 222.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00067560
CAS Number: 65414-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty