Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

L-Tyrosine benzyl ester 95%

Supplier: AMBEED, INC
Warning

A212289-1G A212289-100G A212289-10G A212289-1G A212289-25G A212289-5G
103818-186EA 21 USD InStock
103818-186 103848-884 76825-930 76825-932 77087-350 77087-352 77087-354
L-Tyrosine benzyl ester 95%
L-Tyrosine benzyl ester
Precaución: for Research Use Only


Formula: C₁₆H₁₇NO₃
Boiling Pt: 438.7 °C (760 mm Hg)
Flash Pt: 219.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00191034
CAS Number: 42406-77-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty