Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Phenylpropionic acid 98%

Supplier: AMBEED, INC
Warning

A284136-10G A284136-500G A284136-25G
103809-946EA 93 USD InStock
103809-946 77114-116 77114-118 77305-452
3-Phenylpropionic acid 98%
3-Phenylpropionic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280
Melting Pt: 45…48 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
Merck Index: 13,04805
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty