Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

γ-Aminobutyric acid-t-butyl ester hydrochloride 98%

Supplier: AMBEED, INC
Warning

A147040-1G A147040-5G A147040-100G A147040-10G A147040-25G A147040-5G
103803-728EA 33 USD InStock
103803-728 103841-824 76798-462 76798-464 77299-558 77299-560
γ-Aminobutyric acid-t-butyl ester hydrochloride 98%
gamma-Aminobutyric acid-t-butyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₈ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00270592
CAS Number: 58640-01-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty