Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Imidazo[1,2-a]pyridin-8-ol 95%

Supplier: AMBEED, INC
Warning

A195343-1G A195343-5G A195343-5G A195343-250MG
103816-744EA 18 USD InStock
103816-744 103847-358 77302-458 77671-730
Imidazo[1,2-a]pyridin-8-ol 95%
Imidazo[1,2-a]pyridin-8-ol
Precaución: for Research Use Only


Formula: C₇H₆N₂O
Melting Pt: 154…157 °C
Storage Temperature: Ambient
MDL Number: MFCD06496222
CAS Number: 69214-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty