Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Isochromanone 95%

Supplier: AMBEED, INC
Warning

A116306-1G A116306-5G A116306-100G A116306-10G A116306-250MG A116306-25G A116306-1G A116306-5G
103798-714EA 7 USD InStock
103798-714 103824-574 77026-118 77026-120 77026-122 77026-124 77311-390 77311-392
1-Isochromanone 95%
1-Isochromanone
Precaución: for Research Use Only


Formula: C₉H₈O₂
Boiling Pt: 320.8 °C (760 mm Hg)
Flash Pt: 132.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00799232
CAS Number: 4702-34-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty