Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoindoline hydrochloride 97%

Supplier: AMBEED, INC
Warning

A146060-1G A146060-5G A146060-10G A146060-1G A146060-25G A146060-5G
103803-622EA 5 USD InStock
103803-622 103841-708 76798-420 77299-498 77299-500 77299-502
Isoindoline hydrochloride 97%
Isoindoline hydrochloride
Precaución: for Research Use Only


Formula: C₈H₁₀ClN
Boiling Pt: 239.8 °C (760 mm Hg)
Flash Pt: 98.8 °C
Storage Temperature: Freezer
MDL Number: MFCD03502377
CAS Number: 32372-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty