Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopentyl pentyl phthalate 97%

Supplier: AMBEED, INC
Warning

A200784-1G A200784-5G A200784-250MG A200784-1G
103799-476EA 41 USD InStock
103799-476 103817-238 103847-872 77164-584 77302-720
Isopentyl pentyl phthalate 97%
Isopentyl pentyl phthalate
Precaución: for Research Use Only


Formula: C₁₈H₂₆O₄
MW: 306.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11114532
CAS Number: 776297-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty