Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isopropylboronic acid 97%

Supplier: AMBEED, INC
Warning

A256028-1G A256028-5G A256028-25G A256028-5G A256028-10G A256028-1G
103812-230EA 9 USD InStock
103812-230 103852-090 77091-560 77091-562 77276-432 77304-664
Isopropylboronic acid 97%
Isopropylboronic acid
Precaución: for Research Use Only


Formula: C₃H₉BO₂
MW: 87.91 g/mol
Boiling Pt: 160.4 °C (760 mm Hg)
Melting Pt: 95…100 °C
Storage Temperature: Freezer
MDL Number: MFCD01319021
CAS Number: 80041-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty