Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Isoquinolinol 98%

Supplier: AMBEED, INC
Warning

A126341-1G A126341-100MG A126341-10G A126341-250MG A126341-1G A126341-25G A126341-5G
103800-798EA 31 USD InStock
103800-798 76688-406 76688-408 76688-410 77313-008 77313-010 77313-012
8-Isoquinolinol 98%
8-Isoquinolinol
Precaución: for Research Use Only


Formula: C₉H₇NO
Boiling Pt: 332 °C (760 mm Hg)
Flash Pt: 213.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00661583
CAS Number: 3482-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty