Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Isoquinoline 97%

Supplier: AMBEED, INC
Danger

A359590-1000G A359590-100G A359590-500G A359590-25G
103810-214EA 57 USD InStock
103810-214 76840-418 77115-276 77115-278 77306-702
Isoquinoline 97%
Isoquinoline
Precaución: for Research Use Only


Formula: C₉H₇N
Boiling Pt: 242…243 °C (760 mm Hg)
Melting Pt: 26…28 °C
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD00006898
CAS Number: 119-65-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty