Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Isoxazolecarboxylic acid 96%

Supplier: AMBEED, INC
Warning

A135806-1G A135806-100MG A135806-10G A135806-250MG A135806-1G A135806-5G
103802-224EA 14 USD InStock
103802-224 77162-088 77162-090 77162-092 77315-084 77315-086
3-Isoxazolecarboxylic acid 96%
3-Isoxazolecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₃NO₃
Boiling Pt: 321.2 °C (760 mm Hg)
Melting Pt: 147…153 °C
Flash Pt: 148.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06738796
CAS Number: 3209-71-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty