Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Kartogenin 98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A305493-1G A305493-100MG A305493-10MG A305493-1MG A305493-50MG A305493-5MG A305493-250MG
103814-436EA 589 USD InStock
103814-436 103835-602 76988-862 76988-864 76988-866 76988-868 76988-870 77683-906
Kartogenin 98%, Ambeed.Inc
Kartogenin
Precaución: For research use only.


Formula: C₂₀H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00560510
CAS Number: 4727-31-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty