Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A132651-10MG A132651-25MG A132651-50MG A132651-1MG A132651-10MG
103791-456EA 54 USD InStock
103791-456 103792-124 103792-126 103799-340 77095-024 77298-570
(2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carboxylic acid 95%
(2S,3S)-4-((E)-3-((R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy)-3-oxoprop-1-en-1-yl)-2-(3,4-dihydroxyphenyl)-7-hydroxy-2,3-dihydrobenzofuran-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₂₇H₂₂O₁₂
MW: 538.46 g/mol
Boiling Pt: 862.6 °C (760 mm Hg)
Flash Pt: 291.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03427311
CAS Number: 28831-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty