Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-(+)-Prolinol 97%

Supplier: AMBEED, INC
Warning

A307789-1G A307789-5G A307789-100G A307789-10G A307789-500G A307789-1G A307789-25G A307789-5G
103814-522EA 5 USD InStock
103814-522 103835-698 77290-116 77290-118 77290-120 77305-962 77305-964 77305-966
(S)-(+)-Prolinol 97%
(S)-(+)-Prolinol
Precaución: for Research Use Only


Formula: C₅H₁₁NO
Boiling Pt: 211 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00005255
CAS Number: 23356-96-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty