Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Maleimide 99%

Supplier: AMBEED, INC
Warning

A518248-100G A518248-10G A518248-500G A518248-25G
103839-590EA 7 USD InStock
103839-590 76818-848 76818-850 76818-852 76843-594 77307-700
Maleimide 99%
Maleimide
Precaución: for Research Use Only


Formula: C₄H₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00005494
CAS Number: 541-59-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty