Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl diphenylphosphinite 97%

Supplier: AMBEED, INC
Warning

A208196-5G A208196-10G A208196-1G A208196-25G A208196-5G
103848-548EA 36 USD InStock
103848-548 76721-640 76721-642 77086-854 77086-856
Methyl diphenylphosphinite 97%
Methyl diphenylphosphinite
Precaución: for Research Use Only


Formula: C₁₃H₁₃OP
Boiling Pt: 299.9 °C (760 mm Hg)
Flash Pt: 163.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00048025
CAS Number: 4020-99-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty