Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Nitrothioanisole 95%

Supplier: AMBEED, INC
Warning

A228236-1G A228236-5G A228236-100MG A228236-250MG A228236-1G A228236-5G
103819-376EA 256 USD InStock
103819-376 103850-284 77193-036 77193-038 77303-820 77303-822
2-Nitrothioanisole 95%
2-Nitrothioanisole
Precaución: for Research Use Only


Formula: C₇H₇NO₂S
Boiling Pt: 264.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03094691
CAS Number: 3058-47-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty