Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%

Supplier: AMBEED, INC
Warning

A465033-1G A465033-100MG A465033-250MG
103827-970EA 33 USD InStock
103827-970 76842-952 77196-616 77196-618
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate 97%
Methyl-1,2,3,4-tetrahydroquinoline-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₂
Boiling Pt: 349.4 °C (760 mm Hg)
Melting Pt: 73…76 °C
Flash Pt: 165.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08692534
CAS Number: 177478-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty