Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 1,2,3-triazole-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A186631-1G A186631-5G A186631-10G A186631-250MG A186631-1G A186631-25G A186631-5G
103815-950EA 10 USD InStock
103815-950 103846-536 76998-860 76998-862 77084-208 77084-210 77084-212
Methyl 1,2,3-triazole-4-carboxylate 98%
Methyl 1,2,3-triazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₄H₅N₃O₂
MW: 127.101 g/mol
Boiling Pt: 279.3 °C (760 mm Hg)
Flash Pt: 122.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12912989
CAS Number: 4967-77-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty