Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1H-indazole-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A116194-1G A116194-5G A116194-100G A116194-10G A116194-25G A116194-500G A116194-1G A116194-5G
103798-690EA 5 USD InStock
103798-690 103824-554 77161-008 77161-010 77161-012 77161-014 77311-368 77311-370
Methyl-1H-indazole-3-carboxylate 98%
Methyl 1H-indazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
Boiling Pt: 345.2 °C (760 mm Hg)
Melting Pt: 166…171 °C
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01138133
CAS Number: 43120-28-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty