Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-1H-indazole-7-carboxylate 97%

Supplier: AMBEED, INC
Warning

A119107-1G A119107-10G A119107-25G A119107-1G A119107-5G
103799-164EA 12 USD InStock
103799-164 77233-994 77233-996 77311-836 77311-838
Methyl-1H-indazole-7-carboxylate 97%
Methyl 1H-indazole-7-carboxylate
Precaución: for Research Use Only


Formula: C₉H₈N₂O₂
Boiling Pt: 345.2 °C (760 mm Hg)
Flash Pt: 162.6 °C
Storage Temperature: Ambient
MDL Number: MFCD07371608
CAS Number: 755752-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty