Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(1H-indol-3-yl)acetate 97%

Supplier: AMBEED, INC
Warning

A110895-1G A110895-100G A110895-10G A110895-25G A110895-1G A110895-5G
103797-748EA 10 USD InStock
103797-748 103823-556 76796-834 76796-836 76796-838 77310-352 77310-354
Methyl 2-(1H-indol-3-yl)acetate 97%
Methyl 2-(1H-indol-3-yl)acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Boiling Pt: 341.8 °C (760 mm Hg)
Melting Pt: 50…52 °C
Flash Pt: 160.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005636
CAS Number: 1912-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty