Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate 97%

Supplier: AMBEED, INC
Warning

A338768-1G A338768-100MG A338768-1G
103825-778EA 507 USD InStock
103825-778 103836-752 77195-126 77306-518
Methyl 2-(2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate 97%
Methyl 2-(2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetate
Precaución: for Research Use Only


Formula: C₁₆H₂₃BO₄
Boiling Pt: 385.1±37.0 °C (760 mm Hg)
Flash Pt: 186.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD18732956
CAS Number: 1415960-54-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty