Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-3-(trifluoromethyl)phenylacetate 97%

Supplier: AMBEED, INC
Warning

A234871-1G A234871-5G A234871-100G A234871-10G A234871-1G A234871-25G A234871-5G
103810-986EA 14 USD InStock
103810-986 103850-746 76874-316 76874-318 77089-746 77089-748 77089-750
Methyl-3-(trifluoromethyl)phenylacetate 97%
Methyl-3-(trifluoromethyl)phenylacetate
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O₂
Boiling Pt: 210.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04039762
CAS Number: 62451-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty