Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(3,4-dimethoxyphenyl)acetate 97%

Supplier: AMBEED, INC
Warning

A100282-1G A100282-5G A100282-10G A100282-25G A100282-5G A100282-1G A100282-100G
103795-344EA 13 USD InStock
103795-344 103821-048 76717-482 76717-484 77296-630 77282-468 77666-454
Methyl-2-(3,4-dimethoxyphenyl)acetate 97%
Methyl-2-(3,4-dimethoxyphenyl)acetate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₄
Boiling Pt: 285.4 °C (760 mm Hg)
Flash Pt: 121.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00017206
CAS Number: 15964-79-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty