Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-(bromomethyl)phenylacetate 97%

Supplier: AMBEED, INC
Danger

A182043-1G A182043-25G A182043-100G A182043-100MG A182043-250MG A182043-5G
103807-680EA 24 USD InStock
103807-680 77083-578 77083-580 77287-270 77287-272 77287-274 77287-276
Methyl 4-(bromomethyl)phenylacetate 97%
Methyl 4-(bromomethyl)phenylacetate
Precaución: for Research Use Only


Formula: C₁₀H₁₁BrO₂
Flash Pt: 129.549 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07783030
CAS Number: 7398-42-7
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty