Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-(bromomethyl)-3-nitrobenzoate 95%

Supplier: AMBEED, INC
Danger

A508259-1G A508259-5G
103828-352EA 13 USD InStock
103828-352 103839-454 76843-432 76843-434 77265-768
Methyl-2-(bromomethyl)-3-nitrobenzoate 95%
Methyl 2-(bromomethyl)-3-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₈BrNO₄
MW: 274.07 g/mol
Boiling Pt: 370.9 °C (760 mm Hg)
Melting Pt: 73 °C
Flash Pt: 178.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114315
CAS Number: 98475-07-1
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty