Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(triphenylphosphoranylidene)propanoate 95%

Supplier: AMBEED, INC
Warning

A448349-5G A448349-100MG A448349-1G A448349-5G
103827-776EA 18 USD InStock
103827-776 103838-842 76829-690 76842-654 77307-352
Methyl 2-(triphenylphosphoranylidene)propanoate 95%
Methyl 2-(triphenylphosphoranylidene)propanoate
Precaución: for Research Use Only


Formula: C₂₂H₂₁O₂P
Boiling Pt: 485.8±28.0 °C (760 mm Hg)
Melting Pt: 153…156 °C
Flash Pt: 260.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062441
CAS Number: 2605-68-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty