Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,2-dimethyl-3-oxopropanoate 95%

Supplier: AMBEED, INC
Warning

A100270-1G A100270-5G A100270-100MG A100270-250MG A100270-1G A100270-5G
103795-330EA 150 USD InStock
103795-330 103821-040 76982-122 76982-124 77296-624 77296-626
Methyl 2,2-dimethyl-3-oxopropanoate 95%
Methyl 2,2-dimethyl-3-oxopropanoate
Precaución: for Research Use Only


Formula: C₆H₁₀O₃
Storage Temperature: Freezer
MDL Number: MFCD00082184
CAS Number: 13865-20-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty