Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,3-dichloroquinoxaline-5-carboxylate 95%

Supplier: AMBEED, INC
Warning

A435754-1G A435754-100MG A435754-250MG A435754-50MG
103827-602EA 583 USD InStock
103827-602 76990-560 76990-562 76990-564
Methyl 2,3-dichloroquinoxaline-5-carboxylate 95%
Methyl 2,3-dichloroquinoxaline-5-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₆Cl₂N₂O₂
MW: 257.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27922608
CAS Number: 1643354-85-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty