Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2,4-dihydroxybenzoate 98%

Supplier: AMBEED, INC
Warning

A117300-1000G A117300-10G A117300-25G A117300-500G A117300-5G
103808-170EA 379 USD InStock
103808-170 103824-740 76797-144 76797-146 76797-148 77297-532
Methyl-2,4-dihydroxybenzoate 98%
Methyl-2,4-dihydroxybenzoate
Precaución: for Research Use Only


Formula: C₈H₈O₄
MW: 168.15
Boiling Pt: 300
Melting Pt: 118…121 °C
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00002276
CAS Number: 2150-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty